Anti-HTATIP2 Mouse Polyclonal Antibody
Leverancier: Abnova
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ABNOH00010553-A01EA 258 EUR
ABNOH00010553-A01
Anti-HTATIP2 Mouse Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: HTATIP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human