Anti-POMK Mouse Polyclonal Antibody
ABNOH00084197-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Protein-O-Mannose Kinase
- Antigen symbol:POMK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:84197
- Antigen synonyms:MDDGA12|Probable Inactive Protein Kinase-Like Protein SgK196|MDDGC12|SGK196|Sugen Kinase 196|Protein Kinase-Like Protein SgK196
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: POMK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human