Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?

Anti-FXYD1 Rabbit polyclonal antibody unconjugated

Leverancier: Biorbyt
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: FXYD1 (FXYD domain containing ion transport regulator 1)
Clonality: polyclonal
Conjugation: unconjugated
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

login om uw prijzen en beschikbaarheid te zien
  Onthoud mij op dit apparaat


Expand All / Collapse All
Maat VPE VWR catalogus nummer Hoeveelheid
100 µg 100 µG BIRBORB6893-100
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol FXYD1
Antigen name FXYD domain containing ion transport regulator 1
Reactivity Human, Rat, Mouse
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
Modification phospho-Ser83
Concentratie 0,5 mg/ml
Immunogen KLH conjugated synthetic phosphopeptide derived from human FXYD1 around the phosphorylation site of Ser83.
Moleculair gewicht 10 kDa
Purification purified by affinity chromatography
Form liquid
Opslagbuffer 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
Opslagtemperatuur Cool
Shipping temperature Ice


Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY


Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'X' if unknown for VWR Chemicals. Enter a space if unknown for Merck Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Omschrijving Catalogus nummer Availability Unit Uw prijs Price Per Hoeveelheid