Anti-S100A6 Rabbit Polyclonal Antibody
Leverancier: Biorbyt
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BIRBORB6913-200EA 0 EUR
BIRBORB6913-200 BIRBORB6913-100
Anti-S100A6 Rabbit Polyclonal Antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500
Type: Primary
Antigen: S100A6 (S100 calcium binding protein A6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat