Anti-SIRT1 Rabbit Polyclonal Antibody
Leverancier: Biorbyt
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BIRBORB6959-20EA 0 EUR
BIRBORB6959-20 BIRBORB6959-100
Anti-SIRT1 Rabbit Polyclonal Antibody
Antilichamen
Western Blot: 1:500-1:2000
Type: Primary
Antigen: SIRT1 (sirtuin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now