Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?

Anti-SLC40A1 Rabbit polyclonal antibody unconjugated

Leverancier: Biorbyt
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: SLC40A1 (solute carrier family 40 (iron-regulated transporter), member 1)
Clonality: polyclonal
Conjugation: unconjugated
Host: Rabbit
Isotype: IgG

login om uw prijzen en beschikbaarheid te zien
  Onthoud mij op dit apparaat


Expand All / Collapse All
Maat VPE VWR catalogus nummer Hoeveelheid
100 µg 100 µG BIRBORB6975-100
200 µg 200 µG BIRBORB6975-200
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol Solute carrier family 40, member 1
Antigen name solute carrier family 40 (iron-regulated transporter), member 1
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
Concentratie 0,5 mg/ml - 100 ug (in 200 ul): 0,5 mg/ml- 200 ug (in 400 ul): 0,5 mg/ml
Immunogen KLH conjugated synthetic peptide derived from human SLC40A1/FPN1.
Moleculair gewicht 62 - 69 kDa
Purification purified by Protein A chromatography
Form liquid
Opslagbuffer 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
Opslagtemperatuur Cool
Shipping temperature Ice


Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY


Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'X' if unknown for VWR Chemicals. Enter a space if unknown for Merck Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Omschrijving Catalogus nummer Availability Unit Uw prijs Price Per Hoeveelheid