Anti-TSPAN8 Rabbit polyclonal antibody FITC (Fluorescein)

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BIRBORB7188-100EA 312 EUR
BIRBORB7188-100
Anti-TSPAN8 Rabbit polyclonal antibody FITC (Fluorescein)
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: TSPAN8 (tetraspanin 8)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR