Anti-TSPAN10 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BIRBORB7189-100EA 0 EUR
BIRBORB7189-100
Anti-TSPAN10 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antilichamen
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: TSPAN10 (tetraspanin 10)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR