Anti-EED Rabbit Polyclonal Antibody

Leverancier: Biorbyt
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BIRBORB73985-100EA 0 EUR
BIRBORB73985-100
Anti-EED Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: EED (embryonic ectoderm development)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR