Anti-ABCF2 Mouse Monoclonal Antibody [clone: 2001C1]
Leverancier: Abcam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ABCAAB50807-100EA 510 EUR
ABCAAB50807-100
Anti-ABCF2 Mouse Monoclonal Antibody [clone: 2001C1]
Antilichamen
Type: Primary
Antigen: ABCF2 (ATP-binding cassette, sub-family F (GCN20), member 2)
Clonality: monoclonal
Clone: 2001C1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human