Anti-MFN2 Rabbit polyclonal antibody unconjugated
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ABCAAB50843-100EA 688 EUR
ABCAAB50843-100
Anti-MFN2 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: MFN2 (mitofusin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat