Anti-ANKRD11 Mouse Monoclonal Antibody [clone: 2022C8a]
Leverancier: Abcam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ABCAAB50852-50EA 560 EUR
ABCAAB50852-50
Anti-ANKRD11 Mouse Monoclonal Antibody [clone: 2022C8a]
Antilichamen
Type: Primary
Antigen: ANKRD11 (ankyrin repeat domain 11)
Clonality: monoclonal
Clone: 2022C8a
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now