Anti-MGA Mouse Monoclonal Antibody [clone: MGA6A4H5]
Leverancier: Abcam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ABCAAB50937-50EA 510 EUR
ABCAAB50937-50
Anti-MGA Mouse Monoclonal Antibody [clone: MGA6A4H5]
Antilichamen
Type: Primary
Antigen: MGA (MGA, MAX dimerization protein)
Clonality: monoclonal
Clone: MGA6A4H5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now