Anti-TIMELESS Rabbit Polyclonal Antibody
Leverancier: Abcam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ABCAAB50943-100EA 696 EUR
ABCAAB50943-100
Anti-TIMELESS Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: TIMELESS (timeless circadian clock)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human