Anti-nmt55 / p54nrb Rabbit polyclonal antibody unconjugated
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ABCAAB50952-100EA 688 EUR
ABCAAB50952-100
Anti-nmt55 / p54nrb Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: nmt55 / p54nrb
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human