Anti-VEZF1 Mouse Monoclonal Antibody [clone: ZNF5J141]
Leverancier: Abcam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ABCAAB50970-100EA 510 EUR
ABCAAB50970-100
Anti-VEZF1 Mouse Monoclonal Antibody [clone: ZNF5J141]
Antilichamen
Type: Primary
Antigen: VEZF1 (vascular endothelial zinc finger 1)
Clonality: monoclonal
Clone: ZNF5J141
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat