Anti-BRD1 Mouse monoclonal antibody unconjugated [clone: 2093C5a]
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ABCAAB50989-100EA 672 EUR
ABCAAB50989-100
Anti-BRD1 Mouse monoclonal antibody unconjugated [clone: 2093C5a]
Type: Primary
Antigen: BRD1 (bromodomain containing 1)
Clonality: monoclonal
Clone: 2093C5a
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human