Anti-BTG1 Mouse Monoclonal Antibody [clone: 2095C1a]
Leverancier: Abcam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ABCAAB50991-100EA 530 EUR
ABCAAB50991-100
Anti-BTG1 Mouse Monoclonal Antibody [clone: 2095C1a]
Antilichamen
Type: Primary
Antigen: BTG1 (B-cell translocation gene 1, anti-proliferative)
Clonality: monoclonal
Clone: 2095C1a
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human