Anti-ANXA2 Rabbit polyclonal antibody unconjugated
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ABCAAB90643-200EA 657 EUR
ABCAAB90643-200
Anti-ANXA2 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: ANXA2 (annexin A2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human