Anti-T7 Tag Rabbit Polyclonal Antibody (Agarose)
Leverancier: Abcam
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ABCAAB90665-100EA 550 EUR
ABCAAB90665-100
Anti-T7 Tag Rabbit Polyclonal Antibody (Agarose)
Antilichamen
Type: Primary
Antigen: T7 Tag (T7 tag)
Clonality: polyclonal
Clone:
Conjugation: Agarose
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now