Anti-MT-CO1 Rabbit polyclonal antibody unconjugated
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ABCAAB90668-100EA 672 EUR
ABCAAB90668-100
Anti-MT-CO1 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: MT-CO1 (mitochondrially encoded cytochrome c oxidase I)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human