Anti-IgG Goat Polyclonal Antibody (CF™ 770)
Leverancier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20078.EA 336 EUR
20078. 20078-1
Anti-IgG Goat Polyclonal Antibody (CF™ 770)
Antilichamen
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 770 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Rabbit