Anti-15-PGDH Rabbit Polyclonal Antibody
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PIEAPA116666EA 0 EUR
PIEAPA116666
Anti-15-PGDH Rabbit Polyclonal Antibody
Antilichamen
Suggested positive control: antigen standard for HPGD (transient overexpression lysate), Treated A549 lung carcinoma cell lysates and LoVo colon carcinoma cell lysates. 15-Hydroxyprostaglandin dehydrogenase (15-PGDH) is a novel tumor suppressor in the COX-2 pathway. The protein is found in many normal tissues and appears to be down-regulated in colorectal and lung carcinoma tissue.
Type: Primary
Antigen: 15-PGDH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Mouse