Anti-SCP1 Rabbit Polyclonal Antibody
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PIEAPA116763EA 0 EUR
PIEAPA116763
Anti-SCP1 Rabbit Polyclonal Antibody
Antilichamen
This antibody is not recommended for Immunohistochemistry on paraffin-embedded sections. The synaptonemal complex is a proteinaceous complex that apparently mediates synapses during the zygotene stage and then disintegrates. It is a complex structure that unites homologous chromosomes during the prophase stage of meiosis. It is the result of chromosome pairing interactions and not the cause of them.
Type: Primary
Antigen: SCP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Primates, Rat, Mouse, Human