Anti-P2RX7/P2X7 receptor Goat Polyclonal Antibody
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PIEAPA519165EA 0 EUR
PIEAPA519165
Anti-P2RX7/P2X7 receptor Goat Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: P2RX7/P2X7 receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human