Anti-GLB1 Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))

Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ROCK200-4236EA 3740 EUR
ROCK200-4236
Anti-GLB1 Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Antilichamen
Type: Primary
Antigen: GLB1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR