Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ROCK609-4018EA 333 EUR
ROCK609-4018
Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antilichamen
Secondary Rabbit Anti-IgG F(ab')2 Reacts with Human
Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope: F(ab')2
Host: Rabbit
Isotype: IgG
Reactivity: Human