Anti-IgA/IgG/IgM Goat Polyclonal Antibody (Peroxidase)
Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ROCK610-103-130EA 416 EUR
ROCK610-103-130
Anti-IgA/IgG/IgM Goat Polyclonal Antibody (Peroxidase)
Antilichamen
Suitable for immunoblotting (western or dot blot), ELISA, immunoperoxidase electron microscopy and immunochemistry as well as other peroxidase-antibody based enzymatic assays requiring lot-to-lot consistency.
Recommended Dilutions: ELISA: 1:100000; Western Blot: 1:500 - 1:3000; Immunochemistry: 1:250 - 1:1500;
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgA/IgG/IgM
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgA
Reactivity: Mouse