Anti-IgG gamma Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Leverancier: Rockland Immunochemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ROCK610-4012EA 366 EUR
ROCK610-4012
Anti-IgG gamma Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antilichamen
Secondary Rabbit Anti-IgG (gamma chain) Reacts with Mouse
Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgG γ
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope: γ chain
Host: Rabbit
Isotype: IgG
Reactivity: Mouse