Anti-YARS Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverancier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBI029400EA 400 EUR
USBI029400
Anti-YARS Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antilichamen
Type: Primary
Antigen: YARS (tyrosyl-tRNA synthetase)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR