Anti-NCOA4 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Leverancier: US Biological
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
USBI038836-PEEA 852 EUR
USBI038836-PE
Anti-NCOA4 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antilichamen
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: NCOA4 (nuclear receptor coactivator 4)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human