Anti-ND4L Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
USBI038841-PEEA 551 EUR
USBI038841-PE
Anti-ND4L Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: ND4L
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human