Anti-OR7G1 Rabbit Polyclonal Antibody (Biotin)
Leverancier: US Biological
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
USBI039481-BIOTINEA 852 EUR
USBI039481-BIOTIN
Anti-OR7G1 Rabbit Polyclonal Antibody (Biotin)
Antilichamen
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: OR7G1 (olfactory receptor, family 7, subfamily G, member 1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human