Anti-OR8J3 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBI039490-APEA 852 EUR
USBI039490-AP
Anti-OR8J3 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antilichamen
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: OR8J3 (olfactory receptor, family 8, subfamily J, member 3)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR