Anti-OR8K1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBI039491-HRPEA 852 EUR
USBI039491-HRP
Anti-OR8K1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antilichamen
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: OR8K1 (olfactory receptor, family 8, subfamily K, member 1)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR