Anti-OR9Q1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBI039497-PEEA 852 EUR
USBI039497-PE
Anti-OR9Q1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antilichamen
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: OR9Q1 (olfactory receptor, family 9, subfamily Q, member 1)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR