Anti-ORAI1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBI039498-FITCEA 852 EUR
USBI039498-FITC
Anti-ORAI1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antilichamen
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: ORAI1 (ORAI calcium release-activated calcium modulator 1)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR