Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?

Anti-ORC2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverancier: US Biological
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: ORC2 (origin recognition complex, subunit 2)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human




 

Order

Expand All / Collapse All
Maat VPE VWR catalogus nummer Hoeveelheid
200 µl 200 µl USBI039504-FITC
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Specificaties

Antigen symbol ORC2
Antigen name origin recognition complex, subunit 2
Reactivity Human
Conjugation FITC (Fluorescein Isothiocyanate)
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Amino acid number 207 to 237
Epitope internal
Purification purified by Protein A chromatography
Form liquid
Opslagbuffer PBS, pH 7,2, contains ~0,09% sodium azide
Opslagtemperatuur Cool
Shipping temperature Ice
Tested applications Fluoresence ELISA

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'X' if unknown for VWR Chemicals. Enter a space if unknown for Merck Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Of
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Omschrijving Catalogus nummer Availability Unit Uw prijs Price Per Hoeveelheid