Anti-ORC3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBI039505-HRPEA 852 EUR
USBI039505-HRP
Anti-ORC3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antilichamen
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50

Type: Primary
Antigen: ORC3 (origin recognition complex, subunit 3)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR