Anti-ORMDL3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBI039508-APCEA 852 EUR
USBI039508-APC
Anti-ORMDL3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antilichamen
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: ORMDL3 (ORM1-like 3 (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR