Houd mijn sessie geopend
Eindigt in 
De sessie is verlopen
Uw sessie is verlopen. Voor uw veiligheid bent u uitgelogd.
Wilt u opnieuw inloggen?
Anti-SH3RF3 Rabbit Polyclonal Antibody (Biotin)
Leverancier: US Biological
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: SH3RF3 (SH3 domain containing ring finger 3)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order
Expand All / Collapse All
Maat VPE VWR catalogus nummer Hoeveelheid
200 µl 200 µl USBI041697-BIOTIN
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Specificaties
Antigen symbol SH3RF3
Antigen name SH3 domain containing ring finger 3
Reactivity Mouse
Conjugation Biotin
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Amino acid number 800 to 829
Epitope C-terminal
Purification purified by Protein A chromatography
Form liquid
Opslagbuffer PBS, pH 7,2
Opslagtemperatuur Cool
Shipping temperature Ice
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'X' if unknown for VWR Chemicals. Enter a space if unknown for Merck Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Of
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Omschrijving Catalogus nummer Availability Unit Uw prijs Price Per Hoeveelheid