Anti-TRIM72 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverancier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBI043263-FITCEA 852 EUR
USBI043263-FITC
Anti-TRIM72 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antilichamen
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: TRIM72 (tripartite motif containing 72, E3 ubiquitin protein ligase)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR