Anti-ERAL1 Mouse Polyclonal Antibody

Leverancier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBI126416EA 522 EUR
USBI126416
Anti-ERAL1 Mouse Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: ERAL1 (Era-like 12S mitochondrial rRNA chaperone 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR