Anti-QRSL1 Mouse Polyclonal Antibody

Leverancier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBI132152EA 522 EUR
USBI132152
Anti-QRSL1 Mouse Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: QRSL1 (glutaminyl-tRNA synthase (glutamine-hydrolyzing)-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR