Anti-PBRM1 Rabbit Polyclonal Antibody

Leverancier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBI141930-50EA 358 EUR
USBI141930-50
Anti-PBRM1 Rabbit Polyclonal Antibody
Antilichamen
Recommended dilution: ELISA: 1:100-1:200; Western Blot: 1:50-1:400; Immunochemistry: 1:10-1:500

Type: Primary
Antigen: PBRM1 (polybromo 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR