Anti-TDGF1 Mouse Monoclonal Antibody (Biotin) [clone: 7H257]
Leverancier: United States Biological
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
USBIC7922-10EA 910 EUR
USBIC7922-10
Anti-TDGF1 Mouse Monoclonal Antibody (Biotin) [clone: 7H257]
Antilichamen
Type: Primary
Antigen: TDGF1 (teratocarcinoma-derived growth factor 1)
Clonality: Monoclonal
Clone: 7H257
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human