Anti-GHRL Mouse Monoclonal Antibody [clone: 5E610]
Leverancier: United States Biological
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
USBIG2033-74GEA 1020 EUR
USBIG2033-74G
Anti-GHRL Mouse Monoclonal Antibody [clone: 5E610]
Antilichamen
Type: Primary
Antigen: GHRL (ghrelin/obestatin prepropeptide)
Clonality: monoclonal
Clone: 5E610
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human