Anti-HSF1 Rat Monoclonal Antibody [clone: 0.N.318]

Leverancier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBIH1832-500EA 244 EUR
USBIH1832-500 USBIH1832-250
Anti-HSF1 Rat Monoclonal Antibody [clone: 0.N.318]
Antilichamen
Type: Primary
Antigen: HSF1 (heat shock transcription factor 1)
Clonality: monoclonal
Clone: 0.N.318
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR