Anti-Interleukin 12 p40 Sheep Polyclonal Antibody

Leverancier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBII8434-29BEA 704 EUR
USBII8434-29B
Anti-Interleukin 12 p40 Sheep Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: Interleukin 12 p40
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR