Anti-Mycobacterium tuberculosis Rv2626 Dormant Protein from H37Rv Strain Mouse Monoclonal Antibody [clone: 9B99]

Leverancier: United States Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USBIM9699-64FEA 553 EUR
USBIM9699-64F
Anti-Mycobacterium tuberculosis Rv2626 Dormant Protein from H37Rv Strain Mouse Monoclonal Antibody [clone: 9B99]
Type: Primary
Antigen: Mycobacterium tuberculosis Rv2626 Dormant Protein from H37Rv Strain
Clonality: monoclonal
Clone: 9B99
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR